Assay Details
Gene Symbol: Kcnmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176861.1 | NP_789831.1 | 3 - 4 | 423 | 73 | |
GenBank mRNA | AJ517198.1 | - | 2 - 3 | 307 | 73 | |
AY191836.1 | - | 3 - 4 | 423 | 73 | ||
BC081695.1 | - | 723 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
