Assay Details
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177927.2 | NP_808788.1 | 1 - 2 | 38 | 82 | |
GenBank mRNA | AY216659.1 | - | 1 - 2 | 38 | 82 | |
BC078686.1 | - | 1 - 2 | 26 | 82 | ||
FQ218413.1 | - | 1 - 2 | 38 | 82 | ||
FQ219756.1 | - | 1 - 2 | 37 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY216659.1, BC078686.1, NM_177927.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
