Assay Details
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177927.2 | NP_808788.1 | 3 - 4 | 329 | 58 | |
GenBank mRNA | AY216659.1 | - | 3 - 4 | 329 | 58 | |
BC078686.1 | - | 3 - 4 | 317 | 58 | ||
FQ218413.1 | - | 3 - 4 | 329 | 58 | ||
FQ219756.1 | - | 3 - 4 | 328 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
