Assay Details
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177927.2 | NP_808788.1 | 4 - 5 | 482 | 66 | |
GenBank mRNA | AY216659.1 | - | 4 - 5 | 482 | 66 | |
BC078686.1 | - | 4 - 5 | 470 | 66 | ||
FQ218413.1 | - | 4 - 5 | 482 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
