Assay Details
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177927.2 | NP_808788.1 | 5 - 6 | 690 | 61 | |
GenBank mRNA | AY216659.1 | - | 5 - 6 | 690 | 61 | |
BC078686.1 | - | 5 - 6 | 678 | 61 | ||
FQ218413.1 | - | 5 - 6 | 690 | 61 | ||
FQ219756.1 | - | 5 - 6 | 689 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
