Assay Details
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177927.2 | NP_808788.1 | 7 - 8 | 1043 | 77 | |
GenBank mRNA | AY216659.1 | - | 7 - 8 | 1043 | 77 | |
BC078686.1 | - | 7 - 8 | 1031 | 77 | ||
FQ218413.1 | - | 7 - 8 | 1044 | 77 | ||
FQ219756.1 | - | 7 - 8 | 1043 | 77 | ||
FQ220670.1 | - | 1 - 2 | 178 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
