Assay Details
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182820.1 | NP_877972.1 | 10 - 11 | 1156 | 71 | |
XM_006247588.2 | XP_006247650.1 | 12 - 13 | 1334 | 71 | ||
XM_006247595.2 | XP_006247657.1 | 11 - 12 | 1304 | 71 | ||
GenBank mRNA | AY327409.1 | - | 10 - 11 | 1156 | 71 | |
BC081911.1 | - | 11 - 12 | 1240 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
