Assay Details
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182820.1 | NP_877972.1 | 1 - 2 | 76 | 93 | |
XM_006247586.2 | XP_006247648.1 | 1 - 2 | 141 | 93 | ||
XM_006247588.2 | XP_006247650.1 | 1 - 2 | 140 | 93 | ||
XM_006247595.2 | XP_006247657.1 | 1 - 2 | 139 | 93 | ||
XM_008768359.1 | XP_008766581.1 | 1 - 2 | 141 | 93 | ||
XM_008768360.1 | XP_008766582.1 | 1 - 2 | 141 | 93 | ||
XM_008768361.1 | XP_008766583.1 | 1 - 2 | 140 | 93 | ||
XM_008768362.1 | XP_008766584.1 | 1 - 2 | 140 | 93 | ||
XM_008768363.1 | XP_008766585.1 | 1 - 2 | 140 | 93 | ||
XM_008768364.1 | XP_008766586.1 | 1 - 2 | 140 | 93 | ||
GenBank mRNA | AY327409.1 | - | 1 - 2 | 76 | 93 | |
BC081911.1 | - | 1 - 2 | 75 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY327409.1, BC081911.1, NM_182820.1, XM_006247586.2, XM_006247588.2, XM_006247595.2, XM_008768359.1, XM_008768360.1, XM_008768361.1, XM_008768362.1, XM_008768363.1, XM_008768364.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
