Assay Details
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182820.1 | NP_877972.1 | 2 - 3 | 138 | 114 | |
XM_008768362.1 | XP_008766584.1 | 2 - 3 | 202 | 114 | ||
GenBank mRNA | AY327409.1 | - | 2 - 3 | 138 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY327409.1, NM_182820.1, XM_008768362.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
