Assay Details
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182820.1 | NP_877972.1 | 5 - 6 | 472 | 81 | |
XM_006247586.2 | XP_006247648.1 | 7 - 8 | 651 | 81 | ||
XM_006247588.2 | XP_006247650.1 | 7 - 8 | 650 | 81 | ||
XM_006247595.2 | XP_006247657.1 | 6 - 7 | 620 | 81 | ||
XM_008768359.1 | XP_008766581.1 | 7 - 8 | 651 | 81 | ||
XM_008768360.1 | XP_008766582.1 | 7 - 8 | 627 | 81 | ||
XM_008768361.1 | XP_008766583.1 | 5 - 6 | 563 | 81 | ||
XM_008768362.1 | XP_008766584.1 | 5 - 6 | 539 | 81 | ||
XM_008768363.1 | XP_008766585.1 | 6 - 7 | 621 | 81 | ||
XM_008768364.1 | XP_008766586.1 | 6 - 7 | 597 | 81 | ||
GenBank mRNA | AY327409.1 | - | 5 - 6 | 472 | 81 | |
BC081911.1 | - | 6 - 7 | 556 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
