Assay Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183326.2 | NP_899155.1 | 5 - 6 | 694 | 79 | |
XM_006246123.2 | XP_006246185.1 | 5 - 6 | 686 | 79 | ||
XM_006246124.2 | XP_006246186.1 | 5 - 6 | 519 | 79 | ||
GenBank mRNA | AY574250.1 | - | 4 - 5 | 470 | 79 | |
BC100061.1 | - | 5 - 6 | 694 | 7122007 | 79 | |
FQ211937.1 | - | 5 - 6 | 947 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
