Assay Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033898.1 | NP_001029070.1 | 1 - 2 | 35 | 68 | |
NM_022535.2 | NP_071980.2 | 1 - 2 | 35 | 68 | ||
GenBank mRNA | BC101908.1 | - | 1 - 2 | 35 | 68 | |
CK476875.1 | - | 1 - 2 | 56 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC101908.1, NM_001033898.1, NM_022535.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
