Assay Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033898.1 | NP_001029070.1 | 2 - 3 | 266 | 80 | |
GenBank mRNA | BC101908.1 | - | 2 - 3 | 266 | 80 | |
FQ222101.1 | - | 1 - 2 | 109 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
