Assay Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033898.1 | NP_001029070.1 | 4 - 5 | 626 | 135 | |
NM_022535.2 | NP_071980.2 | 4 - 5 | 626 | 135 | ||
GenBank mRNA | BC101908.1 | - | 4 - 5 | 626 | 135 | |
CK476875.1 | - | 4 - 5 | 647 | 135 | ||
FQ222101.1 | - | 3 - 4 | 469 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
