Assay Details
Gene Symbol: Nnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106819.1 | NP_001100289.1 | 1 - 2 | 347 | 102 | |
XM_008766195.1 | XP_008764417.1 | 1 - 2 | 572 | 102 | ||
GenBank mRNA | FQ218983.1 | - | 1 - 2 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
