Assay Details
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107241.1 | NP_001100711.1 | 3 - 4 | 437 | 99 | |
XM_006251539.2 | XP_006251601.1 | 4 - 5 | 576 | 99 | ||
XM_006251540.2 | XP_006251602.1 | 3 - 4 | 464 | 99 | ||
XM_006251541.2 | XP_006251603.1 | 3 - 4 | 458 | 99 | ||
XM_006251542.2 | XP_006251604.1 | 3 - 4 | 491 | 99 | ||
GenBank mRNA | BC099114.1 | - | 3 - 4 | 397 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107241.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
