Assay Details
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107241.1 | NP_001100711.1 | 5 - 6 | 653 | 84 | |
XM_006251539.2 | XP_006251601.1 | 6 - 7 | 788 | 84 | ||
XM_006251540.2 | XP_006251602.1 | 5 - 6 | 676 | 84 | ||
XM_006251541.2 | XP_006251603.1 | 5 - 6 | 670 | 84 | ||
XM_006251542.2 | XP_006251604.1 | 5 - 6 | 703 | 84 | ||
GenBank mRNA | BC099114.1 | - | 6 - 7 | 692 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
