Assay Details
Gene Symbol: Hrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BF387663.1 | - | 1 | 148 | 67 | |
D83697.1 | - | 2 | 5122 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BF387663.1 | - | 1 | 148 | 67 | |
D83697.1 | - | 2 | 5122 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |