Assay Details
Gene Symbol: Ppp1r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138521.1 | NP_612530.1 | 6 - 7 | 991 | 80 | |
XM_006247344.2 | XP_006247406.1 | 3 - 4 | 1489 | 80 | ||
GenBank mRNA | AY601872.1 | - | 6 - 7 | 991 | 80 | |
BC078954.1 | - | 6 - 7 | 921 | 7110316 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
