Assay Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082477.2 | NP_001075946.2 | 2 - 3 | 204 | 67 | |
NM_001082478.1 | NP_001075947.1 | 2 - 3 | 501 | 67 | ||
NM_001082479.1 | NP_001075948.1 | 2 - 3 | 501 | 67 | ||
NM_178866.4 | NP_849197.3 | 2 - 3 | 204 | 67 | ||
XM_006241191.2 | XP_006241253.1 | 3 - 4 | 345 | 67 | ||
XM_006241192.1 | XP_006241254.1 | 3 - 4 | 259 | 67 | ||
XM_006241193.2 | XP_006241255.1 | 3 - 4 | 344 | 67 | ||
XM_006241194.2 | XP_006241256.1 | 2 - 3 | 383 | 67 | ||
GenBank mRNA | BC086374.1 | - | 2 - 3 | 204 | 7300903 | 67 |
D00698.1 | - | 2 - 3 | 185 | 67 | ||
FQ209824.1 | - | 3 - 4 | 333 | 67 | ||
FQ210560.1 | - | 2 - 3 | 217 | 67 | ||
FQ210688.1 | - | 2 - 3 | 217 | 67 | ||
FQ220385.1 | - | 2 - 3 | 525 | 67 | ||
FQ225388.1 | - | 2 - 3 | 217 | 67 | ||
FQ225437.1 | - | 2 - 3 | 217 | 67 | ||
FQ231951.1 | - | 2 - 3 | 217 | 67 | ||
M15480.1 | - | 2 - 3 | 248 | 67 | ||
M15481.1 | - | 2 - 3 | 936 | 67 | ||
X06043.1 | - | 1 - 2 | 86 | 67 | ||
X06107.1 | - | 2 - 3 | 479 | 67 | ||
X06108.1 | - | 3 - 4 | 293 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
