Assay Details
Gene Symbol: Gadd45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024127.2 | NP_077041.2 | 3 - 4 | 533 | 127 | |
GenBank mRNA | AB102787.1 | - | 3 - 4 | 383 | 127 | |
AB102788.1 | - | 3 - 4 | 383 | 127 | ||
BC081795.1 | - | 3 - 4 | 533 | 127 | ||
L32591.1 | - | 3 - 4 | 401 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
