Assay Details
Gene Symbol: Il3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246226.2 | XP_006246288.1 | 2 - 3 | 213 | 78 | |
GenBank mRNA | U81493.1 | - | 2 - 3 | 228 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
