Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297634.1 | NP_001284563.1 | 4 - 5 | 2278 | 74 | |
XM_021085696.1 | XP_020941355.1 | 3059 | 74 | |||
XM_021085698.1 | XP_020941357.1 | 4464 | 74 | |||
XM_021085699.1 | XP_020941358.1 | 2606 | 74 | |||
GenBank mRNA | KJ652909.1 | - | 4 - 5 | 2278 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
