Assay Details
Gene Symbol: HSPA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005661695.3 | XP_005661752.1 | 648 | 87 | ||
GenBank mRNA | AK394133.1 | - | 3 - 4 | 310 | 87 | |
AK396015.1 | - | 3 - 4 | 310 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
