Assay Details
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243317.1 | NP_001230246.1 | 4 - 5 | 382 | 125 | |
GenBank mRNA | AK349860.1 | - | 4 - 5 | 382 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
