Assay Details
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013996829.2 | XP_013852283.1 | 1540 | 72 | ||
XM_021089536.1 | XP_020945195.1 | 1791 | 72 | |||
XM_021089538.1 | XP_020945197.1 | 1544 | 72 | |||
GenBank mRNA | AK397273.1 | - | 6 - 7 | 981 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
