Assay Details
Gene Symbol: FKBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038000.2 | NP_001033089.1 | 1 - 2 | 190 | 91 | |
GenBank mRNA | AK231994.1 | - | 1 - 2 | 130 | 91 | |
AK235743.1 | - | 1 - 2 | 129 | 91 | ||
AK235766.1 | - | 1 - 2 | 188 | 91 | ||
AK389279.1 | - | 1 - 2 | 130 | 91 | ||
AY610330.1 | - | 1 - 2 | 169 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
