Assay Details
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013982543.2 | XP_013837997.1 | 1220 | 143 | ||
XM_021070833.1 | XP_020926492.1 | 1594 | 143 | |||
GenBank mRNA | AF109646.1 | - | 3 - 4 | 418 | 143 | |
AK393460.1 | - | 3 - 4 | 503 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() |
