Assay Details
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308246.1 | NP_001295175.1 | 16 - 17 | 2206 | 75 | |
XM_003130205.5 | XP_003130253.2 | 2538 | 75 | |||
GenBank mRNA | AF403245.1 | - | 2 - 3 | 250 | 75 | |
AY825267.1 | - | 10 - 11 | 1401 | 75 | ||
KP233220.1 | - | 16 - 17 | 2206 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
