Assay Details
Gene Symbol: TMEFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021075732.1 | XP_020931391.1 | 1801 | 79 | ||
GenBank mRNA | AK390041.1 | - | 2 - 3 | 695 | 79 | |
AK400149.1 | - | 2 - 3 | 79 | |||
AY609989.1 | - | 2 - 3 | 375 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
