Assay Details
Gene Symbol: DEGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244121.1 | NP_001231050.1 | 2 | 339 | 81 | |
XM_005653768.3 | XP_005653825.1 | 364 | 81 | |||
GenBank mRNA | AK236774.1 | - | 2 | 341 | 81 | |
AY609708.1 | - | 2 | 320 | 81 | ||
AY632774.1 | - | 1 | 44 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
