Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162401.1 | NP_001155873.1 | 1 | 407 | 53 | |
XM_005654915.3 | XP_005654972.1 | 582 | 53 | |||
XM_005654916.3 | XP_005654973.1 | 675 | 53 | |||
XM_005654917.3 | XP_005654974.1 | 816 | 53 | |||
XM_005654918.3 | XP_005654975.1 | 662 | 53 | |||
GenBank mRNA | EF183524.1 | - | 1 | 147 | 53 | |
EF546799.1 | - | 1 | 407 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
