Assay Details
Gene Symbol: UBE2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078674.1 | NP_001072142.1 | 2 - 3 | 89 | 76 | |
GenBank mRNA | AK234981.1 | - | 2 - 3 | 276 | 76 | |
DQ972956.1 | - | 2 - 3 | 89 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
