Assay Details
Gene Symbol: UBE2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078674.1 | NP_001072142.1 | 6 - 7 | 402 | 92 | |
XM_021072207.1 | XP_020927866.1 | 589 | 92 | |||
GenBank mRNA | AK234981.1 | - | 6 - 7 | 589 | 92 | |
DQ972956.1 | - | 6 - 7 | 402 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
