Assay Details
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097517.1 | NP_001090986.1 | 2 - 3 | 425 | 145 | |
XM_021068172.1 | XP_020923831.1 | 324 | 145 | |||
GenBank mRNA | AK234353.1 | - | 2 - 3 | 425 | 145 | |
EF486868.1 | - | 2 - 3 | 425 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
