Assay Details
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098591.1 | NP_001092061.1 | 2 - 3 | 210 | 79 | |
GenBank mRNA | AB271947.1 | - | 2 - 3 | 210 | 79 | |
AK400460.1 | - | 2 - 3 | 248 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
