Assay Details
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098598.1 | NP_001092068.1 | 2 - 3 | 153 | 114 | |
GenBank mRNA | AK231505.1 | - | 2 - 3 | 134 | 114 | |
AK234485.1 | - | 2 - 3 | 234 | 114 | ||
AK234557.1 | - | 2 - 3 | 130 | 114 | ||
AK239428.1 | - | 2 - 3 | 130 | 114 | ||
AK347722.1 | - | 2 - 3 | 132 | 114 | ||
AK393088.1 | - | 2 - 3 | 297 | 114 | ||
AK393867.1 | - | 2 - 3 | 266 | 114 | ||
AK394469.1 | - | 2 - 3 | 130 | 114 | ||
AK394975.1 | - | 2 - 3 | 130 | 114 | ||
AK395376.1 | - | 2 - 3 | 348 | 114 | ||
AK395674.1 | - | 2 - 3 | 255 | 114 | ||
AK398383.1 | - | 2 - 3 | 130 | 114 | ||
AK399331.1 | - | 2 - 3 | 132 | 114 | ||
AK400321.1 | - | 2 - 3 | 134 | 114 | ||
AK400933.1 | - | 2 - 3 | 132 | 114 | ||
AK400997.1 | - | 2 - 3 | 130 | 114 | ||
AK401004.1 | - | 2 - 3 | 132 | 114 | ||
DQ316319.1 | - | 2 - 3 | 153 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
