Assay Details
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098598.1 | NP_001092068.1 | 4 - 5 | 410 | 95 | |
GenBank mRNA | AK239428.1 | - | 4 - 5 | 387 | 95 | |
AK393088.1 | - | 4 - 5 | 554 | 95 | ||
AK393867.1 | - | 4 - 5 | 523 | 95 | ||
AK394469.1 | - | 4 - 5 | 387 | 95 | ||
AK395376.1 | - | 4 - 5 | 605 | 95 | ||
AK395674.1 | - | 4 - 5 | 512 | 95 | ||
AK398383.1 | - | 4 - 5 | 387 | 95 | ||
AK398853.1 | - | 4 - 5 | 389 | 95 | ||
AK399331.1 | - | 4 - 5 | 389 | 95 | ||
AK400321.1 | - | 4 - 5 | 391 | 95 | ||
AK400933.1 | - | 4 - 5 | 389 | 95 | ||
AK400997.1 | - | 4 - 5 | 387 | 95 | ||
AK401004.1 | - | 4 - 5 | 389 | 95 | ||
DQ316319.1 | - | 4 - 5 | 410 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
