Assay Details
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101815.1 | NP_001095285.1 | 3 - 4 | 478 | 67 | |
GenBank mRNA | AK233128.1 | - | 3 - 4 | 448 | 67 | |
AK346370.1 | - | 3 - 4 | 538 | 67 | ||
AK394195.1 | - | 3 - 4 | 558 | 67 | ||
AY349422.1 | - | 3 - 4 | 467 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
