Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112689.1 | NP_001106159.2 | 3 - 4 | 251 | 62 | |
XM_005669744.3 | XP_005669801.1 | 386 | 62 | |||
XM_013981921.1 | XP_013837375.1 | 284 | 62 | |||
GenBank mRNA | AK232875.1 | - | 3 - 4 | 263 | 62 | |
EF374062.1 | - | 3 - 4 | 251 | 62 | ||
EU395789.1 | - | 2 - 3 | 215 | 62 | ||
JF923469.1 | - | 2 - 3 | 205 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
