Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112689.1 | NP_001106159.2 | 4 - 5 | 411 | 87 | |
XM_005669744.3 | XP_005669801.1 | 546 | 87 | |||
XM_005669745.3 | XP_005669802.1 | 461 | 87 | |||
XM_013981921.1 | XP_013837375.1 | 444 | 87 | |||
XM_013981922.2 | XP_013837376.1 | 365 | 87 | |||
GenBank mRNA | AK232875.1 | - | 4 - 5 | 423 | 87 | |
AK390817.1 | - | 4 - 5 | 547 | 87 | ||
EF374062.1 | - | 4 - 5 | 411 | 87 | ||
JF923469.1 | - | 3 - 4 | 365 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
