Assay Details
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113039.2 | NP_001106510.2 | 2 - 3 | 514 | 84 | |
GenBank mRNA | AB078418.1 | - | 2 - 3 | 293 | 84 | |
AB188301.2 | - | 2 - 3 | 385 | 84 | ||
AJ628065.1 | - | 2 - 3 | 272 | 84 | ||
AK230585.1 | - | 2 - 3 | 363 | 84 | ||
AK396573.1 | - | 2 - 3 | 514 | 84 | ||
AY535422.1 | - | 2 - 3 | 266 | 84 | ||
EU716413.1 | - | 2 - 3 | 266 | 84 | ||
FJ555286.1 | - | 2 - 3 | 266 | 84 | ||
FJ555287.1 | - | 2 - 3 | 266 | 84 | ||
GQ304754.1 | - | 2 - 3 | 266 | 84 | ||
GU723684.1 | - | 2 - 3 | 266 | 84 | ||
KF460453.1 | - | 2 - 3 | 307 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
