Assay Details
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113217.1 | NP_001106688.1 | 3 - 4 | 342 | 78 | |
GenBank mRNA | AK343747.1 | - | 3 - 4 | 327 | 78 | |
AK343823.1 | - | 3 - 4 | 318 | 78 | ||
AK351256.1 | - | 3 - 4 | 319 | 78 | ||
AY735454.1 | - | 3 - 4 | 342 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
