Assay Details
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113697.1 | NP_001107169.1 | 2 - 3 | 401 | 85 | |
XM_013985516.2 | XP_013840970.1 | 333 | 85 | |||
GenBank mRNA | AK348355.1 | - | 2 - 3 | 266 | 85 | |
AY609503.1 | - | 3 - 4 | 358 | 85 |
Gene Symbol: LOC100525472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666531.3 | XP_005666588.1 | 241 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
