Assay Details
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113697.1 | NP_001107169.1 | 5 - 6 | 663 | 109 | |
XM_013985516.2 | XP_013840970.1 | 595 | 109 | |||
GenBank mRNA | AK348355.1 | - | 5 - 6 | 528 | 109 | |
AK349578.1 | - | 5 - 6 | 585 | 109 | ||
AY609503.1 | - | 6 - 7 | 620 | 109 |
Gene Symbol: LOC100525472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666531.3 | XP_005666588.1 | 503 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
