Assay Details
Gene Symbol: SALL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114673.1 | NP_001108145.1 | 2 - 3 | 2445 | 84 | |
XM_021077202.1 | XP_020932861.1 | 2558 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
