Assay Details
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122995.2 | NP_001116467.1 | 4 - 5 | 619 | 111 | |
NM_001301326.1 | NP_001288255.1 | 4 - 5 | 616 | 111 | ||
GenBank mRNA | AK392493.1 | - | 4 - 5 | 616 | 111 | |
BP172930.1 | - | 4 - 5 | 619 | 111 | ||
CK454400.1 | - | 3 - 4 | 435 | 111 | ||
EU545196.1 | - | 4 - 5 | 449 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
