Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123113.1 | NP_001116585.1 | 2 - 3 | 392 | 78 | |
GenBank mRNA | AK232994.1 | - | 2 - 3 | 551 | 78 | |
JX861095.1 | - | 2 - 3 | 392 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
