Assay Details
Gene Symbol: RPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213940.1 | NP_999105.1 | 5 - 6 | 411 | 78 | |
GenBank mRNA | AB000911.1 | - | 5 - 6 | 411 | 78 | |
AK344436.1 | - | 5 - 6 | 425 | 78 | ||
AY610361.1 | - | 5 - 6 | 417 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
