Assay Details
Gene Symbol: DAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213944.1 | NP_999109.1 | 2 - 3 | 444 | 103 | |
GenBank mRNA | AK349909.1 | - | 2 - 3 | 452 | 103 | |
AK391622.1 | - | 2 - 3 | 454 | 103 | ||
AY609509.1 | - | 2 - 3 | 492 | 103 | ||
D86562.1 | - | 2 - 3 | 444 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
